Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 1,2-Dioctanoyl-sn-glycerol

Catalog No. 31750510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-750-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-750-510MG Supplier MilliporeSigma™ Supplier No. 31750510MG
Add to Cart
Add to Cart

Cell-permeable activator of protein kinase C (PKC).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Description 97% by NMR
Target Proteins Protein kinase C
CAS 60514-48-9
Molecular Formula C19H36O5
Formula Weight 344.6g/mol
Solubility DMSO or Chloroform
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.