Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 11beta-HSD1 Inhibitor, 10j

Catalog No. 38558110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-558-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-558-110MG Supplier MilliporeSigma™ Supplier No. 38558110MG
Add to Cart
Add to Cart

The 11beta-HSD1 Inhibitor, 10j, also referenced under CAS 1009373-58-3, controls the biological activity of 11beta-HSD1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cell permeable, spirocyclic urea that acts as a selective inhibitor of 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) (IC50 of 1.6nM and 5.3 nM) in enzyme and human adipocyte assays, respectively. Consisting of 2 enantiomers, 10j2 (R Isomer) is shown to be ˜ 2-3X more potent than 10j1 (S Isomer), in both aforementioned assays. 11β-HSD1 activity in the presence of 10j2 (10mg/kg in animals p.o.) results in an ˜ 80% reduction in the concentration of plasma cortisol over vehicle treated controls.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
CAS 1009373-58-3
Molecular Formula C26H33BrN2O3
Formula Weight 501.46g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.