Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 1-Aminocyclopropane-1-carboxylic Acid

Catalog No. 149101250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-1250MG 250 mg
14-910-11GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-910-1250MG Supplier MilliporeSigma™ Supplier No. 149101250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An agonist for the glycine modulatory site of the N-methyl-D-aspartate (NMDA) receptor complex.

An agonist for the glycine modulatory site of the N-methyl-D-aspartate (NMDA) receptor complex. Mimics the effects of glycine on the NMDA receptor ion channel and blocks 3H-glycine binding to rat forebrain membrane (IC50 = 38 nM). Blocks NMDA-induced release of arachidonic acid (IC50 = 131μM) in cerebellar granule cells. Reduces NMDA-induced cell death in hippocampal neurons.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins Glycine modulatory site of the N-methyl-D-aspartate (NMDA) receptor complex
CAS 22059-21-8
Molecular Formula C4H7NO2
Formula Weight 101.1g/mol
Solubility H2O
Form Crystalline Solid
Quantity 250 mg
Purity 98% by TLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.