Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AMPK Inhibitor, Compound C

Catalog No. 1712605MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-126-05MG 5 mg
17-126-01MG 1 mg
17-126-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-126-05MG Supplier MilliporeSigma™ Supplier No. 1712605MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Product Type Inhibitor
Target Proteins AMPK
CAS 866405-64-3
Molecular Formula C24H25N5O
Formula Weight 399.5g/mol
Solubility DMSO, 100 mM HCl, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.