Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α1-Antichymotrypsin, Human Plasma

Catalog No. 1781961MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-819-61MG 1 mg
17-819-6100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-819-61MG Supplier MilliporeSigma™ Supplier No. 1781961MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acute-phase protein present at ∽45mg/100mL in plasma.

Acute-phase protein present at ∽45mg/100mL in plasma. Specific inhibitor of chymotrypsin-like serine proteinases. Molecular weight ranges from 65,000-68,000 daltons.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 150 mM NaCl, 20 mM Tris-HCl, pH 7.4.
Target Proteins Chymotrypsin-like serine proteases
CAS 141176-92-3
Solubility Aqueous buffers
Form Solid
Quantity 1 mg
Purity 95% by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.