Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α1-Antichymotrypsin, Human Plasma

Catalog No. p-7060028 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
100 μg

Acute-phase protein present at ∽45mg/100mL in plasma.

Acute-phase protein present at ∽45mg/100mL in plasma. Specific inhibitor of chymotrypsin-like serine proteinases. Molecular weight ranges from 65,000-68,000 daltons.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 150 mM NaCl, 20 mM Tris-HCl, pH 7.4.
Target Proteins Chymotrypsin-like serine proteases
CAS 141176-92-3
Solubility Aqueous buffers
Form Solid
Quantity 100 μg
Purity 95% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.