Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 17-DMAG

Catalog No. 100069500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-006-9500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-006-9500UG Supplier MilliporeSigma™ Supplier No. 100069500UG
Add to Cart
Add to Cart

A potent antitumor analog of 17-AAG that binds to the ATPase site of human Hsp90α with high affinity and displays excellent bioavailability and aqueous solubility.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Purple
Description 95% by TLC
Target Proteins 17-DMAG
CAS 467214-21-7
Molecular Formula C32H48N4O8·HCl
Formula Weight 653.2g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.