Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2,3 cis-Piperidine dicarboxylic acid

Catalog No. 5050510001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-505-10001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-505-10001 Supplier MilliporeSigma™ Supplier No. 5050510001
Add to Cart
Add to Cart

A non-specific antagonist of NMDA, AMPA and kainate ionotropic receptors and a partial agonist for NMDA receptors.

A non-specific antagonist of NMDA, AMPA and kainate ionotropic receptors and a partial agonist for NMDA receptors. Used in blocking general excitatory synaptic transmissions.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Molecular Formula C7H11NO4
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.