Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2',5'-Dideoxyadenosine

Catalog No. 2881041MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-810-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-810-41MG Supplier MilliporeSigma™ Supplier No. 2881041MG
Add to Cart
Add to Cart

Cell-permeable, non-competitive adenylate cyclase inhibitor (IC50 = 3 μM), that binds to the adenosine P1 binding site.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins adenylate cyclase
CAS 6698-26-6
Molecular Formula C10H13N5O2
Formula Weight 235.2g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.