Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ p-Aminophenylmercuric Acetate

Catalog No. 164610700MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
700 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-461-0700MG 700 mg
16-461-01SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-461-0700MG Supplier MilliporeSigma™ Supplier No. 164610700MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An organomercurial agent used for the activation of latent MMPs in vitro.

An organomercurial agent used for the activation of latent MMPs in vitro. Acts by facilitating the loss of the enzyme propeptide domain through an autolytic cleavage known as the cysteine switch.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins Activation of latent MMPs in vitro
CAS 6283-24-5
Molecular Formula C8H9HgNO2
Formula Weight 351.8g/mol
Solubility DMSO or 0.1 N NaOH
Form Solid
Quantity 700 mg
Purity 90% by titration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.