Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2-Chloro-2'-deoxyadenosine

Catalog No. 22046710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-046-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-046-710MG Supplier MilliporeSigma™ Supplier No. 22046710MG
Add to Cart
Add to Cart

A deoxyadenosine analog that displays strong antileukemic and immunosuppressive activity.

A deoxyadenosine analog that displays strong antileukemic and immunosuppressive activity. Also induces apoptosis in chronic lymphocytic leukemia (CLL) cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Antileukemic and immunosuppressive activity
CAS 4291-63-8
Molecular Formula C10H12ClN5O3
Formula Weight 285.7g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.