Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2-N-Hexadecanoylamino-4-nitrophenylphosphorylcholine

Catalog No. 5048590001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-485-90001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-485-90001 Supplier MilliporeSigma™ Supplier No. 5048590001
Add to Cart
Add to Cart

Synthetic chromogenic analog of sphingomyelin.

Synthetic chromogenic analog of sphingomyelin. Useful substrate for selectively measuring the activity of acid sphingomyelinase.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by TLC
Formula Weight 575.7g/mol
Solubility MeOH
Content And Storage Protect from light
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.