Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 3-Isobutyl-1-methylxanthine

Catalog No. 410957250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-095-7250MG 250 mg
41-095-71GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-095-7250MG Supplier MilliporeSigma™ Supplier No. 410957250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC50 = 2-50 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Inhibitor
Target Proteins cAMP and cGMP phosphodiesterases
CAS 28822-58-4
Molecular Formula C10H14N4O2
Formula Weight 222.2g/mol
Solubility DMSO, EtOH, or MeOH
Form Solid
Quantity 250 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.