Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Q-VD-OPh, Non-O-methylated

Catalog No. 5514765MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-147-65MG 5 mg
55-147-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-147-65MG Supplier MilliporeSigma™ Supplier No. 5514765MG
Add to Cart
Add to Cart

InSolution™ Q-VD-OPh, Non-O-methylated, is a cell-permeable, irreversible, broad-spectrum inhibitor of caspases (IC50 = 50, 100, 430, and <25 nM for caspase-1,-8, -9, and -3, respectively).

  • Packaged Under Inert Gas
  • Peptide Sequence: Q-Val-Asp-CH2-OPh
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 10 mM (1mg/195μl) solution in DMSO.
Target Proteins caspase-1
Molecular Formula C26H25F2N3O6
Formula Weight 513.8g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg
Purity 90% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.