Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 5,6-Dichloro-1-β-D-ribofuranosylbenzimidazole

Catalog No. 28789150MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-789-150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-789-150MG Supplier MilliporeSigma™ Supplier No. 28789150MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, ATP-competitive, and specific inhibitor of casein kinase II (IC50 = 6 μM).

A potent, ATP-competitive, and specific inhibitor of casein kinase II (IC50 = 6μM). Reported to inhibit insulin-stimulated nuclear and cytosolic p70S6 kinase in CHO cells. Accentuates premature termination of transcription. Has been used to inhibit RNA polymerase II transcription which may be dependent on casein kinase II. Inhibits germinal vesicle breakdown in bovine oocytes. Induces G1/S cell cycle arrest.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins casein kinase II
CAS 53-85-0
Molecular Formula C12H12Cl2N2O4
Formula Weight 319.2g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.