Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 5,7-Dichlorokynurenic Acid, Sodium Salt

Catalog No. 5048410001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-484-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-484-10001 Supplier MilliporeSigma™ Supplier No. 5048410001
Add to Cart
Add to Cart

A very potent, water-soluble antagonist of the NMDA receptors complex acting at the strychnine-insensitive glycine binding site (Ki = 79 nM

A very potent, water-soluble antagonist of the NMDA receptors complex acting at the strychnine-insensitive glycine binding site (Ki = 79nM vs. [3H]-glycine).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins NMDA receptors
Molecular Formula C10H4Cl2NNaO3
Formula Weight 280.04g/mol
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.