Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PAR-2 Agonist I

Catalog No. 5391091MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-910-91MG 1 mg
53-910-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-910-91MG Supplier MilliporeSigma™ Supplier No. 5391091MG
Add to Cart
Add to Cart

The PAR-2 Agonist I controls the biological activity of PAR-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: 2-Furoyl-Leu-Ile-Gly-Arg-Leu-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins PAR-2
Molecular Formula C31H53N9O7
Formula Weight 663.8g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.