Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Simvastatin, Sodium Salt

Catalog No. 5670215MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-702-15MG 5 mg
56-702-150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-702-15MG Supplier MilliporeSigma™ Supplier No. 5670215MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simvastatin, Sodium Salt is a carboxylate form of Simvastatin. Active in whole cells and in cell-free preparations. An inhibitor of HMG-CoA reductase.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Target Proteins 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase
Molecular Formula C25H39O6·Na
Formula Weight 458.6g/mol
Solubility DMSO, EtOH, or H
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Purity 97% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.