Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ACC2 Inhibitor, CD-017-0191

Catalog No. 10455110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-455-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-455-110MG Supplier MilliporeSigma™ Supplier No. 10455110MG
Add to Cart
Add to Cart

The ACC2 Inhibitor, CD-017-0191 controls the biological activity of ACC2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

A cell-permeable and selective inhibitor of human Acetyl-CoA carboxylase 2 (ACC2) (IC50 = 2.8μM), which does not inhibit human ACC1 at all at concentrations up to 100μM, in vitro . It is also shown to inhibit the growth of yeast (IC50 = 3.8μM), in which the native ACC1 gene is replaced with the human ACC2 strain; but not with the human ACC1 strain at concentrations ≤ 100μM. This compound is not demonstrated to be toxic to human cells in culture at concentrations up to 10μM.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Molecular Formula C20H19N3O3S
Formula Weight 381.45g/mol
Solubility DMSO
Content And Storage Toxicity: Standard Handling, Store at 2°C to 8°C, Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.