Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Acetyl-Pepstatin

Catalog No. 1101751MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-017-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-017-51MG Supplier MilliporeSigma™ Supplier No. 1101751MG
Add to Cart
Add to Cart

An aspartyl protease inhibitor that acts as an effective inhibitor of HIV-1 proteinase (Ki = 20nM at pH 4.7) and HIV-2 proteinase (Ki = 5nM at pH 4.7).

An aspartyl protease inhibitor that acts as an effective inhibitor of HIV-1 proteinase (Ki = 20nM at pH 4.7) and HIV-2 proteinase (Ki = 5nM at pH 4.7). Acts as a substrate-analog inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins HIV-1 proteinase
CAS 28575-34-0
Molecular Formula C31H57N5O9
Formula Weight 643.8g/mol
Solubility 50% acetic acid
Content And Storage Toxicity: Standard Handling, Store at -20°C, Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.