Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ALLN

Catalog No. 2087195MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-871-95MG 5 mg
20-871-925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-871-95MG Supplier MilliporeSigma™ Supplier No. 2087195MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ALLN, CAS 110044-82-1, is a cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).

  • Peptide Sequence: N-Acetyl-Leu-Leu-Nle-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Product Type Protease Inhibitor
Target Proteins Calpain-1
CAS 110044-82-1
Molecular Formula C20H37N3O4
Formula Weight 383.5g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.