Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calphostin C, Cladosporium cladosporioides

Catalog No. 20872550UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-872-550UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-872-550UG Supplier MilliporeSigma™ Supplier No. 20872550UG

missing translation for 'validMainframeMsg'

A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein's regulatory domain by competing at the binding site of diacylglycerol and phorbol esters.

A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein’s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters. Does not compete with Ca2+ or phospholipids. At higher concentrations inhibits myosin light chain kinase (IC50 > 5μM), protein kinase A (IC50 > 50μM), protein kinase G (IC50 > 25μM), and p60v- src (IC50 > 50μM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Requires brief exposure to light for activation.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Brown
Target Proteins PKC
CAS 121263-19-2
Molecular Formula C44H38O14
Formula Weight 790.8g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Lyophilized
Quantity 50 μg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.