Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-9 Substrate I, Fluorogenic

Catalog No. 2187655MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-876-55MG 5 mg
21-876-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-876-55MG Supplier MilliporeSigma™ Supplier No. 2187655MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A fluorogenic substrate for caspase-9 (ICE-LAP6, Mch6).

Peptide Sequence: Ac-Leu-Glu-His-Asp-AFC
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins A fluorogenic substrate for caspase-9
Molecular Formula C33H38F3N7O11
Formula Weight 765.7g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.