Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AdaAhx3L3VS

Catalog No. 114802250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-480-2250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-480-2250UG Supplier MilliporeSigma™ Supplier No. 114802250UG
Add to Cart
Add to Cart

A potent, covalent, cell-permeable, irreversible inhibitor of chymotrypsin-like (50-100 nM), trypsin-like (1-5 μM), and PGPH (0.5-1 μM) activities of the 20S proteasome.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Target Proteins chymotrypsin-like activity of the 20S proteasome
CAS 389064-25-9
Molecular Formula C50H88N6O8S
Formula Weight 932.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 250 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.