Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenophostin A, Hexasodium Salt

Catalog No. 11550050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-550-050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-550-050UG Supplier MilliporeSigma™ Supplier No. 11550050UG
Add to Cart
Add to Cart

Adenophostin A, Hexasodium Salt, is a potent agonist of type 1 inositol 1,4,5-trisphosphate receptor (InsP3R; EC50 = 10.9nM). Exhibits about 50- to 100-fold greater affinity for IP3R than IP3.

One of the most potent known agonist of type 1 inositol 1,4,5-trisphosphate receptor (InsP3R; EC50 = 10.9nM). Exhibits about 50- to 100-fold greater affinity for IP3R than IP3. Stimulates Ca2+ release from Ins(1,4,5)P3-sensitive Ca2+ stores in microsomal preparations, permeabilized cells, and lipid vesicles containing purified InsP3R (EC50 = 14.7nM for Ca2+ release from permeabilized hepatocytes). Does not bind to Ins(1,3,4,5)P4 binding sites and is resistant to enzymatic degradation.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by HPLC
Target Proteins Type 1 inositol 1,4,5-trisphosphate receptor (InsP3R)
Molecular Formula C16H20N5O18P3·6Na
Formula Weight 801.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.