Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, 8-(4-Chlorophenylthio)-2'-O-Methyl-, Sodium Salt
Shop All MilliporeSigma ProductsDescription
- Potent, cell-permeable, and specific activator of the exchange protein activated by cyclic AMP in vitro and in vivo
- Does not have any effect on protein kinase A (PKA)
- A useful tool to discriminate between EPAC and PKA
Specifications
Specifications
| Content And Storage | Store at −20°C |
| Formula Weight | 507.8g/mol |
| Molecular Formula | C17H16ClN5O6PS·Na |
| Purity | ≥98% by HPLC |
| Quantity | 5 μmol |
| Synonym | 8-CPT-2′-O-Me-cAMP, Na |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?