Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3 Substrate II, Fluorogenic

Catalog No. 2354255MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-542-55MG 5 mg
23-542-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-542-55MG Supplier MilliporeSigma™ Supplier No. 2354255MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic caspase-3 substrate.

Peptide Sequence: Ac-Asp-Glu-Val-Asp-AMC
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Fluorogenic caspase-3 substrate
Molecular Formula C30H37N5O13
Formula Weight 675.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.