Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3 Inhibitor I

Catalog No. 2354201MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-542-01MG 1 mg
23-542-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-542-01MG Supplier MilliporeSigma™ Supplier No. 2354201MG
Add to Cart
Add to Cart

The Caspase-3 Inhibitor I, also referenced under CAS 169332-60-9, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Ac-Asp-Glu-Val-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Capase-3, capase-6, capase-7, capase-8, capase-10
CAS 169332-60-9
Molecular Formula C20H30N4O11
Formula Weight 502.5g/mol
Solubility DMSO or H2O
Form Solid
Quantity 1 mg
Purity 90% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.