Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6-Benzoyl-, Sodium Salt

Catalog No. 11680210UMO Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-680-210UMO 10 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-680-210UMO Supplier MilliporeSigma™ Supplier No. 11680210UMOL
Add to Cart
Add to Cart

A potent cell-permeable activator of cAMP-dependent protein kinase (PKA).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins PKA
CAS 30275-80-0
Molecular Formula C17H15N5O7P·Na
Formula Weight 455.3g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 10 μmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.