Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Colchicine, Colchicum autumnale

Catalog No. 2341151GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-411-51GM 1 g
23-411-55GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-411-51GM Supplier MilliporeSigma™ Supplier No. 2341151GM
Add to Cart
Add to Cart

Colchicine, Colchicum autumnale, CAS 64-86-8, is an inhibitor of mitosis that disrupts microtubules and inhibits tubulin polymerization. Induces apoptosis in PC12 and cerebellar granule cells.

Inhibitor of mitosis that is useful in cell division studies. Disrupts microtubules and inhibits tubulin polymerization. Induces apoptosis in PC12 cells and in cerebellar granule cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow or Off-white
Target Proteins Inhibitor of mitosis
CAS 64-86-8
Molecular Formula C22H25NO6
Formula Weight 399.4g/mol
Solubility EtOH or H2O
Content And Storage Protect from light
Form Powder
Quantity 1 g
Purity 94% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.