Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ C5a Receptor Antagonist, W-54011

Catalog No. 2344155MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-441-55MG 5 mg
23-441-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-441-55MG Supplier MilliporeSigma™ Supplier No. 2344155MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The C5a Receptor Antagonist, W-54011, also referenced under CAS 405098-33-1, controls the biological activity of C5a Receptor.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins C5a Receptor
CAS 405098-33-1
Molecular Formula C30H36N2O2·HCl
Formula Weight 493.1g/mol
Solubility DMSO or MeOH
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.