Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DNA-PK Inhibitor V
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Yellow |
| Target Proteins | DNA-PK |
| CAS | 404009-46-7 |
| Molecular Formula | C17H17NO3 |
| Formula Weight | 283.3g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 5 mg |
| Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction