Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclopamine, V. californicum

Catalog No. 2398035MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-980-35MG 5 mg
23-980-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-980-35MG Supplier MilliporeSigma™ Supplier No. 2398035MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopamine, V. californicum, CAS 4449-51-8, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonizes Sonic Hedgehog signaling through direct interaction with Smo.

A cell-permeable steroidal alkaloid and cholesterol mimic that displays both teratogenic and antitumor activities. It disrupts cholesterol bio-synthesis and specifically antagonizes shh (Sonic Hedgehog) signaling pathway through direct interaction with Smo (smoothened), a distant relative of G-protein-coupled receptors. A valuable tool for studying the involvement of shh signaling in the development of various tumors. Tomatidine (Cat. No. 614350) can be used as a negative control. Also available as a 10mm solution in Ethanol (Cat. No. 239806).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Hh signaling in Shh-light2 assay
CAS 4449-51-8
Molecular Formula C27H41NO2
Formula Weight 411.6g/mol
Solubility DMSO, EtOH, or DMF
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 97% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.