Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 5'-Diphosphate, Potassium Salt

Catalog No. 1171051GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-710-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-710-51GM Supplier MilliporeSigma™ Supplier No. 1171051GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For biochemical applications

TRUSTED_SUSTAINABILITY

Specifications

Format White solid
Product Type Adenosine 5′-Diphosphate, Potassium Salt
Content And Storage 2°C to 8°C
Purity ≥98% by enzymatic assay (dry basis)
Quantity 1 g

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.