Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GSK-3 Inhibitor XIII

Catalog No. 3615555MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-155-55MG 5 mg
36-155-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-155-55MG Supplier MilliporeSigma™ Supplier No. 3615555MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The GSK-3 Inhibitor XIII, also referenced under CAS 404828-08-6, controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

An aminopyrazole compound that acts as a potent and ATP-binding site inhibitor of GSK-3 with a Ki of 24 nM.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins GSK-3
CAS 404828-08-6
Molecular Formula C18H15N5
Formula Weight 301.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.