Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GM 6001, Negative Control

Catalog No. 3642105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-421-05MG 5 mg
36-421-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-421-05MG Supplier MilliporeSigma™ Supplier No. 3642105MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A useful negative control for the MMP inhibitor GM 6001.

A useful negative control for the MMP inhibitor GM 6001 (Cat. No. 364205).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins A useful negative control for the MMP inhibitor GM 6001
CAS 877381-76-5
Molecular Formula C23H34N4O4
Formula Weight 430.5g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.