Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GM 6001, Negative Control

Catalog No. p-7060082 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

A useful negative control for the MMP inhibitor GM 6001.

A useful negative control for the MMP inhibitor GM 6001 (Cat. No. 364205).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins A useful negative control for the MMP inhibitor GM 6001
CAS 877381-76-5
Molecular Formula C23H34N4O4
Formula Weight 430.5g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.