Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gö 6976

Catalog No. 365250500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-525-0500UG 500 μg
36-525-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-525-0500UG Supplier MilliporeSigma™ Supplier No. 365250500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC50 = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC50 = 2.3 nM) and βI (IC50 = 6.2 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins PKC
CAS 136194-77-9
Molecular Formula C24H18N4O
Formula Weight 378.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.