Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, N6,O2'-Dibutyryl-, Sodium Salt

Catalog No. 2874525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-745-25MG 25 mg
28-745-100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-745-25MG Supplier MilliporeSigma™ Supplier No. 2874525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Dibutyryl-cAMP, CAS 16980-89-5, is a cell-permeable cAMP analog that preferentially activates PKA.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins PKA
CAS 16980-89-5
Molecular Formula C18H23N5O8P·Na
Formula Weight 491.4g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 25 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.