Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GSK-3β Inhibitor XII, TWS119

Catalog No. p-7060079 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-155-41MG 1 mg
36-155-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-155-41MG Supplier MilliporeSigma™ Supplier No. 3615541MG
Only null left
Add to Cart
Add to Cart

The GSK-3β Inhibitor XII, TWS119, also referenced under CAS 601514-19-6, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Target Proteins GSK-3β
CAS 601514-19-6
Molecular Formula C18H14N4O2
Formula Weight 318.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.