Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ KT5720

Catalog No. p-7060092 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-032-050UG 50 μg
42-032-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-032-050UG Supplier MilliporeSigma™ Supplier No. 42032050UG
Only null left
Add to Cart
Add to Cart

A potent, specific, cell-permeable, reversible and ATP-competitive inhibitor of protein kinase A (Ki = 56 nM) that is prepared by a chemical modification of K-252a.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins PKA
CAS 108068-98-0
Molecular Formula C32H31N3O5
Formula Weight 537.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 μg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.