Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Diethyl Pyrocarbonate

Catalog No. 298711100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-871-1100GM 100 g
29-871-125GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-871-1100GM Supplier MilliporeSigma™ Supplier No. 298711100GM
Add to Cart
Add to Cart

Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles.

Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles. Useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Also inhibits platelet-activating factor acetyl hydrolase. Has been used as a probe for the topography of 5.8S rRNA in yeast ribosomes.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Target Proteins Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle
CAS 1609-47-8
Molecular Formula C6H10O5
Formula Weight 162.1g/mol
Solubility CHCl3 or EtOH
Content And Storage Protect from light
Form Liquid
Quantity 100 g
Purity 97% by titration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.