Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Eg5 Inhibitor III, Dimethylenastron

Catalog No. 3246225MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-462-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-462-25MG Supplier MilliporeSigma™ Supplier No. 3246225MG
Add to Cart
Add to Cart

The Eg5 Inhibitor III, Dimethylenastron, also referenced under CAS 863774-58-7, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Membrane applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Target Proteins Microtubule-stimulated ATPase activity of the mitotic motor
CAS 863774-58-7
Molecular Formula C16H18N2O2S
Formula Weight 302.4g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.