Learn More
MilliporeSigma™ Calbiochem™ Advanced Glycation End product-BSA
Catalog No. 12180010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
12-180-010MG 10 mg
1 options
Catalog No. 12-180-010MG Supplier MilliporeSigma™ Supplier No. 12180010MG
Add to Cart
Add to Cart
Prepared by reacting BSA with glycoaldehyde under sterile conditions.
  • Reported to induce apoptosis in cultured human umbilical vein endothelial cells
  • Inhibits nitric oxide synthase activity in proximal tubular epithelial cells
  • Advanced glycation end products their receptors have been implicated in pathogenesis of diabetes, induction of proinflammatory cytokines, stimulation of smooth muscle proliferation, fibronectin production
TRUSTED_SUSTAINABILITY
Specifications
Description BSA
Formulation In sterile-filtered PBS
Quantity 10 mg