Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AEBSF, HCl ≥99% by HPLC
GSA_VA

Catalog No. 5325860001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-258-60001 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-258-60001 Supplier MilliporeSigma™ Supplier No. 5325860001
Add to Cart
Add to Cart

Specific irreversible inhibitor of serine proteases

Specific irreversible inhibitor of serine proteases. Reacts covalently with a serine at the active site. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. Stable, nontoxic alternative to PMSF (Cat. No. 52332) and DFP (Cat. No. 30967). Used at the same molar concentration (0.1-1.0 mM) as PMSF for most applications. Exhibits low inhibitory activity towards proteinase K. A material with ≥97% purity by HPLC (Cat. No. 101500) and its InSolution™ is also available (Cat. No. 508436).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins Serine proteases
Molecular Formula C8H10FNO2S·HCl
Solubility H2O
Content And Storage Protect from Light, Protect from Moisture
Form Powder
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.