Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Autophagy Inhibitor, 3-MA

Catalog No. 18949050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-949-050MG 50 mg
18-949-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-949-050MG Supplier MilliporeSigma™ Supplier No. 18949050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Autophagy Inhibitor, 3-MA, CAS 5142-23-4, is a cell-permeable autophagic sequestration blocker. Acts as an inhibitor of III PI3-Kinase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White to Faint Yellow
CAS 5142-23-4
Molecular Formula C6H7N5
Formula Weight 149.2g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Powder
Quantity 50 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.