Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (±)-Bay K 8644

Catalog No. 1968781MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-687-81MG 1 mg
19-687-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-687-81MG Supplier MilliporeSigma™ Supplier No. 1968781MG
Add to Cart
Add to Cart

Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types.

Synthetic dihydropyridine derivative that acts as an active Ca2+ slow channel agonist in neuroendocrine, muscle, thyroid and other cell types. Prolongs single channel open time without affecting the close time. An L-type Ca2+ channel agonist. Composed of two optical isomers. The (-)-enantiomer has strong vasoconstrictive, positive inotropic, and Ca2+ agonistic properties, whereas the (+)-enantiomer has weakly vasodilating, negative inotropic, and Ca2+ antagonistic properties. The net effect of the racemic mix is that of the negative enantiomer. Promotes β-cell proliferation/regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins L-type Ca2+ channel
CAS 93468-89-4
Molecular Formula C16H15F3N2O4
Formula Weight 356.3g/mol
Solubility EtOH or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.