Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AG 1295

Catalog No. 6585505MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-855-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-855-05MG Supplier MilliporeSigma™ Supplier No. 6585505MG
Add to Cart
Add to Cart

A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC50 = 500nM) and PDGF-dependent DNA synthesis (IC50 = 2.5μM) in Swiss/3T3 cells.

A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC50 = 500nM) and PDGF-dependent DNA synthesis (IC50 = 2.5μM) in Swiss/3T3 cells. Has no effect on EGF receptor autophosphorylation and shows only a weak effect on EGF- or insulin-stimulated DNA synthesis. A potent blocker of smooth muscle cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins PDGFR kinase
CAS 71897-07-9
Molecular Formula C16H14N2
Formula Weight 234.3g/mol
Solubility DMSO or CH2Cl2
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.