Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AG 1478

Catalog No. 6585525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-855-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-855-25MG Supplier MilliporeSigma™ Supplier No. 6585525MG
Add to Cart
Add to Cart

A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins Epidermal growth factor receptor kinase
CAS 175178-82-2
Molecular Formula C16H14ClN3O2
Formula Weight 315.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.