Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AhR Agonist V, VAF347

Catalog No. 18269010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-269-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-269-010MG Supplier MilliporeSigma™ Supplier No. 18269010MG
Add to Cart
Add to Cart

The AhR Agonist V, VAF347 controls the biological activity of AhR. This small molecule/inhibitor is primarily used for Biochemicals applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Molecular Formula C17H11ClF3N3
Formula Weight 349.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.